Product Details

SNP ID
rs736413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:45827467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGATGATTTAGGGTTGGGTCCC[A/G]CTGAGACAGTGGCCAGACCCAAGTG
Phenotype
MIM: 615068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPG5 PubMed Links

Gene Details

Gene
EPG5
Gene Name
ectopic P-granules autophagy protein 5 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020964.2 Intron NP_066015.2
Gene
SIGLEC15
Gene Name
sialic acid binding Ig like lectin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213602.2 Intron NP_998767.1

View Full Product Details