Product Details

SNP ID
rs480104
Assay Type
Validated
NCBI dbSNP Submissions
37
Location
Chr.1:160831256 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCCATATCCTCTCCAGACTAGGAA[C/G]TGTTCTATAGAGACTCCTGTGCCGT
Phenotype
MIM: 605554 MIM: 600684
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD244 PubMed Links

Gene Details

Gene
CD244
Gene Name
CD244 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166663.1 1429 UTR 3 NP_001160135.1
NM_001166664.1 1429 UTR 3 NP_001160136.1
NM_016382.3 1429 UTR 3 NP_057466.1
XM_011509621.2 1429 Intron XP_011507923.1
XM_011509622.2 1429 Intron XP_011507924.1
XM_011509623.2 1429 UTR 3 XP_011507925.1
Gene
LY9
Gene Name
lymphocyte antigen 9
There are no transcripts associated with this gene.

View Full Product Details