Product Details

SNP ID
rs3891248
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:127737893 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGTGCCTTGGATTTTGGCAAATTGT[A/T]TTCCTCACCGCCACCTCCCGCGGCT
Phenotype
MIM: 190080
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CASC11 PubMed Links

Gene Details

Gene
CASC11
Gene Name
cancer susceptibility candidate 11 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MYC
Gene Name
v-myc avian myelocytomatosis viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002467.4 Intron NP_002458.2

View Full Product Details