Product Details
- SNP ID
-
rs260219
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:110051541 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCAGATTTCTGTCACGGTTATTA[C/G]CAGTCTCTGCTTATTAAGGTAATGA
- Phenotype
-
MIM: 604582
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AKAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139972484] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKAP2
- Gene Name
- A-kinase anchoring protein 2
- Gene
- PALM2-AKAP2
- Gene Name
- PALM2-AKAP2 readthrough
View Full Product Details