Product Details

SNP ID
rs425729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:117524401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGCATTTGTTTTAAAGAAGTGG[G/T]TCCTGGTATACACTAGATGTCCGTT
Phenotype
MIM: 610237
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MED30 PubMed Links

Gene Details

Gene
MED30
Gene Name
mediator complex subunit 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282986.1 Intron NP_001269915.1
NM_080651.3 Intron NP_542382.1
XM_011517360.2 Intron XP_011515662.1

View Full Product Details