Product Details

SNP ID
rs199396
Assay Type
Functionally Tested
NCBI dbSNP Submissions
11
Location
Chr.3:38138992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGAGCTGCTTACCAAGCTGGGC[C/T]GCGACGACGTGCTGCTGGAGCTGGG
Phenotype
MIM: 604054 MIM: 602170
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACAA1 PubMed Links

Gene Details

Gene
ACAA1
Gene Name
acetyl-CoA acyltransferase 1
There are no transcripts associated with this gene.

Gene
MYD88
Gene Name
myeloid differentiation primary response 88
There are no transcripts associated with this gene.

View Full Product Details