Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286431.1 | 10484 | UTR 3 | NP_001273360.1 | ||
NM_001286432.1 | 10484 | UTR 3 | NP_001273361.1 | ||
NM_002912.4 | 10484 | UTR 3 | NP_002903.3 | ||
XM_011536028.1 | 10484 | UTR 3 | XP_011534330.1 | ||
XM_011536029.2 | 10484 | UTR 3 | XP_011534331.1 | ||
XM_011536030.2 | 10484 | UTR 3 | XP_011534332.1 | ||
XM_011536031.2 | 10484 | UTR 3 | XP_011534333.1 | ||
XM_011536032.2 | 10484 | UTR 3 | XP_011534334.1 | ||
XM_011536036.2 | 10484 | Intron | XP_011534338.1 | ||
XM_017011152.1 | 10484 | UTR 3 | XP_016866641.1 | ||
XM_017011153.1 | 10484 | UTR 3 | XP_016866642.1 | ||
XM_017011154.1 | 10484 | UTR 3 | XP_016866643.1 | ||
XM_017011155.1 | 10484 | Intron | XP_016866644.1 |