Product Details

SNP ID
rs544021
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:66548190 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATGACAGGAGATGACCCCAAGGGC[G/C]GAGAGGCTCCGCTGACTCGGCTGGG
Phenotype
MIM: 102574
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
ACTN3 PubMed Links

Gene Details

Gene
ACTN3
Gene Name
actinin alpha 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104.3 Intron NP_001095.2
NM_001258371.2 Intron NP_001245300.2
Gene
ZDHHC24
Gene Name
zinc finger DHHC-type containing 24
There are no transcripts associated with this gene.

View Full Product Details