Product Details

SNP ID
rs157252
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:16470226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACCCATGGACGGCTTGAATAAG[A/C]TGATGGGTTAAGAAAGGTTTCTGCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CROCCP3 PubMed Links

Gene Details

Gene
CROCCP3
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details