Product Details
- SNP ID
-
rs599634
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:50045950 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATACGTTCAGTGGATACAATATTA[C/T]AAATAGTCATACAACAATTCTGTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB112
PubMed Links
Gene Details
- Gene
- DEFB112
- Gene Name
- defensin beta 112
View Full Product Details