Product Details

SNP ID
rs422907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33590881 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTGAGGGCAGAGGGCTGGGCTC[A/G]GGTCAAGGCTAGAATGGTGAGAGCC
Phenotype
MIM: 609495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGNBP1 PubMed Links

Gene Details

Gene
GGNBP1
Gene Name
gametogenetin binding protein 1 (pseudogene)
There are no transcripts associated with this gene.

Gene
LINC00336
Gene Name
long intergenic non-protein coding RNA 336
There are no transcripts associated with this gene.

View Full Product Details