Product Details

SNP ID
rs273242
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:78623278 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGATAATCTTGTCTTGTACTTTA[C/T]CTTAGAGGAAAAGCTGTCAGTTTTT
Phenotype
MIM: 613975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI44L PubMed Links

Gene Details

Gene
IFI44L
Gene Name
interferon induced protein 44 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006820.3 Intron NP_006811.2
XM_005270391.3 Intron XP_005270448.1
XM_005270392.4 Intron XP_005270449.1
XM_005270393.4 Intron XP_005270450.1
XM_006710304.3 Intron XP_006710367.1
XM_011540539.2 Intron XP_011538841.1
XM_017000120.1 Intron XP_016855609.1

View Full Product Details