Product Details
- SNP ID
-
rs344071
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:156826795 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCAGGAAAGTCTTAGGATGTGAACT[C/T]CCGCCTTCACCTGCCCCTGACCTGC
- Phenotype
-
MIM: 613536
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LEKR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62273904,rs6767106] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LEKR1
- Gene Name
- leucine, glutamate and lysine rich 1
- Gene
- LINC00886
- Gene Name
- long intergenic non-protein coding RNA 886
There are no transcripts associated with this gene.
View Full Product Details