Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003559.4 | 5129 | UTR 3 | NP_003550.1 | ||
XM_011525326.2 | 5129 | UTR 3 | XP_011523628.1 | ||
XM_011525327.2 | 5129 | UTR 3 | XP_011523629.1 | ||
XM_011525329.2 | 5129 | Intron | XP_011523631.1 | ||
XM_011525330.2 | 5129 | UTR 3 | XP_011523632.1 | ||
XM_017025196.1 | 5129 | UTR 3 | XP_016880685.1 | ||
XM_017025197.1 | 5129 | UTR 3 | XP_016880686.1 | ||
XM_017025198.1 | 5129 | UTR 3 | XP_016880687.1 | ||
XM_017025199.1 | 5129 | UTR 3 | XP_016880688.1 |