Product Details

SNP ID
rs597778
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:43143337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAGGAGAAGCAAGGAACTGTCC[A/G]TAAAGAAGCAGTCTAAGGAAATAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM183A PubMed Links

Gene Details

Gene
FAM183A
Gene Name
family with sequence similarity 183 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101376.2 379 Intron NP_001094846.2
XM_005270875.4 379 Intron XP_005270932.1
XM_005270876.4 379 UTR 5 XP_005270933.1
XM_017001322.1 379 UTR 5 XP_016856811.1

View Full Product Details