Product Details

SNP ID
rs235256
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:12236175 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATGCTTCTCCGCAGGGTAGAA[A/C]AATGCTTTTTCTTTTGTAACTGGTG
Phenotype
MIM: 608877
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VPS13D PubMed Links

Gene Details

Gene
VPS13D
Gene Name
vacuolar protein sorting 13 homolog D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015378.3 Intron NP_056193.2
NM_018156.3 Intron NP_060626.2

View Full Product Details