Product Details

SNP ID
rs654092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125889796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCTGGAAAAACGGATTATCCCC[A/C]ACACACAGGGCATAATAAGGATGCT
Phenotype
MIM: 610693 MIM: 616283
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HYLS1 PubMed Links

Gene Details

Gene
HYLS1
Gene Name
HYLS1, centriolar and ciliogenesis associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134793.1 Intron NP_001128265.1
NM_145014.2 Intron NP_659451.1
XM_005271430.2 Intron XP_005271487.1
XM_006718777.3 Intron XP_006718840.1
XM_011542659.2 Intron XP_011540961.1
XM_017017320.1 Intron XP_016872809.1
XM_017017321.1 Intron XP_016872810.1
Gene
PUS3
Gene Name
pseudouridylate synthase 3
There are no transcripts associated with this gene.

View Full Product Details