Product Details

SNP ID
rs30985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2226663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTATCCCTTTGTGTAAACAGG[C/T]ACTGTCCCTGACACTCAGCCGTTTG
Phenotype
MIM: 602622 MIM: 603022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L2 PubMed Links

Gene Details

Gene
DNASE1L2
Gene Name
deoxyribonuclease I-like 2
There are no transcripts associated with this gene.

Gene
E4F1
Gene Name
E4F transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288776.1 Intron NP_001275705.1
NM_001288778.1 Intron NP_001275707.1
NM_004424.4 Intron NP_004415.3
XM_005255155.1 Intron XP_005255212.1
XM_005255156.1 Intron XP_005255213.1
XM_006720858.1 Intron XP_006720921.1
XM_011522402.1 Intron XP_011520704.1
XM_011522403.1 Intron XP_011520705.1
XM_017023010.1 Intron XP_016878499.1
XM_017023011.1 Intron XP_016878500.1

View Full Product Details