Product Details

SNP ID
rs219294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60092943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGTATCCATCATTTTTCACTTA[C/T]TCATTTTTTACTATTTTTAGGTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCNX4 PubMed Links

Gene Details

Gene
PCNX4
Gene Name
pecanex homolog 4 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022495.5 Intron NP_071940.4
XM_005268009.2 Intron XP_005268066.1

View Full Product Details