Product Details

SNP ID
rs33935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20539422 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACAATCATTGAGCAAAGCTTA[A/G]ATCTTGCCTTTGTTTCTTGTTAGTC
Phenotype
MIM: 603984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF737 PubMed Links

Gene Details

Gene
ZNF737
Gene Name
zinc finger protein 737
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159293.1 6961 UTR 3 NP_001152765.1
XM_005259697.3 6961 Intron XP_005259754.1
XM_006722597.3 6961 Intron XP_006722660.1
XM_011527598.2 6961 Intron XP_011525900.1
XM_011527599.2 6961 Intron XP_011525901.1
XM_017026116.1 6961 Intron XP_016881605.1
XM_017026117.1 6961 Intron XP_016881606.1

View Full Product Details