Product Details

SNP ID
rs36136
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:65594123 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAACATTCATGCTATTATCTTCT[C/T]TGTCTATAATGTTCTTCCTTTCACA
Phenotype
MIM: 601747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPWD1 PubMed Links

Gene Details

Gene
PPWD1
Gene Name
peptidylprolyl isomerase domain and WD repeat containing 1
There are no transcripts associated with this gene.

Gene
TRIM23
Gene Name
tripartite motif containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001656.3 Intron NP_001647.1
NM_033227.2 Intron NP_150230.1
NM_033228.2 Intron NP_150231.1
XM_005248497.3 Intron XP_005248554.1
XM_017009444.1 Intron XP_016864933.1

View Full Product Details