Product Details

SNP ID
rs159872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58036081 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGGACTCCCTTTCTATGACACT[A/C]ACGCATCATCCTGCATCCCACGTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN1 PubMed Links

Gene Details

Gene
ZSCAN1
Gene Name
zinc finger and SCAN domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182572.3 1339 Intron NP_872378.3
XM_006723149.2 1339 UTR 5 XP_006723212.1
XM_017026609.1 1339 UTR 5 XP_016882098.1
XM_017026610.1 1339 Intron XP_016882099.1

View Full Product Details