Product Details

SNP ID
rs173174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6740015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGGAAACTGAGGCACAGGGTTG[A/C]GAATGAGGCGGGCAGAAGGAATCCT
Phenotype
MIM: 604504
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR108 PubMed Links

Gene Details

Gene
GPR108
Gene Name
G protein-coupled receptor 108
There are no transcripts associated with this gene.

Gene
MIR6791
Gene Name
microRNA 6791
There are no transcripts associated with this gene.

Gene
TRIP10
Gene Name
thyroid hormone receptor interactor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288962.1 Intron NP_001275891.1
NM_001288963.1 Intron NP_001275892.1
NM_004240.3 Intron NP_004231.1
XM_005259683.2 Intron XP_005259740.1
XM_006722940.1 Intron XP_006723003.1

View Full Product Details