Product Details

SNP ID
rs292263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89839771 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAAAGTTTAGTATAAAGAATTA[C/T]TGGTCGGGTTCCGTGGCTCAGGCCT
Phenotype
MIM: 606880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP8AP2 PubMed Links

Gene Details

Gene
CASP8AP2
Gene Name
caspase 8 associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137667.1 Intron NP_001131139.1
NM_001137668.1 Intron NP_001131140.1
NM_012115.3 Intron NP_036247.1

View Full Product Details