Product Details

SNP ID
rs457665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89836591 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATTTCAGAGGTAGCATCAGTA[A/G]ATCAGTAGTCTGTTAGGGAATGTTA
Phenotype
MIM: 606880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP8AP2 PubMed Links

Gene Details

Gene
CASP8AP2
Gene Name
caspase 8 associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137667.1 Intron NP_001131139.1
NM_001137668.1 Intron NP_001131140.1
NM_012115.3 Intron NP_036247.1
Gene
MDN1
Gene Name
midasin AAA ATPase 1
There are no transcripts associated with this gene.

View Full Product Details