Product Details

SNP ID
rs121917839
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177993032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCTTCCGTTGCTTAGCTCTGC[G/A]GTTCTGGAACCAGACCTGAGAAGGG
Phenotype
MIM: 601538
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
PROP1 PubMed Links

Gene Details

Gene
PROP1
Gene Name
PROP paired-like homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006261.4 667 Missense Mutation CGC,TGC R120C NP_006252.3

View Full Product Details