Product Details

SNP ID
rs34468236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:19950342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGTTCTAAAAGTTTCCCATTT[A/G]TTAATAACATAATTTTTCTCAAAAC
Phenotype
MIM: 179512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFHB PubMed Links

Gene Details

Gene
EFHB
Gene Name
EF-hand domain family member B
There are no transcripts associated with this gene.

Gene
RAB5A
Gene Name
RAB5A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292048.1 Intron NP_001278977.1
NM_004162.4 Intron NP_004153.2

View Full Product Details