Product Details

SNP ID
rs4617087
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:76393180 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTCACAGCTGCACCACGCGGCGCTC[A/C]CCAGGCCCCGCCCCATTTTCCACTG
Phenotype
MIM: 607639 MIM: 182889
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SSC4D PubMed Links

Gene Details

Gene
SSC4D
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080744.1 Intron NP_542782.1
XM_017011750.1 Intron XP_016867239.1
Gene
ZP3
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
There are no transcripts associated with this gene.

View Full Product Details