Product Details
- SNP ID
-
rs12508354
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:133967912 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTAGTCTTATGAAATATGAAAAT[G/T]TGTTTAGAATTGTGGATGTCCCAAT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PABPC4L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77100830] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PABPC4L
- Gene Name
- poly(A) binding protein cytoplasmic 4 like
View Full Product Details