Product Details

SNP ID
rs11992321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23195783 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGAAACACAATCTTTTAGTAT[G/T]AAATTAACCTTTGATTCCTGGGTGG
Phenotype
MIM: 603611
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF10A PubMed Links

Gene Details

Gene
TNFRSF10A
Gene Name
TNF receptor superfamily member 10a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003844.3 Intron NP_003835.3

View Full Product Details