Product Details

SNP ID
rs10180455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3381255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAATACGTGCCACCTAGATTTC[C/T]ACTACACACCCTCGTGCCCTTATGG
Phenotype
MIM: 614139 MIM: 608998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC12 PubMed Links

Gene Details

Gene
TRAPPC12
Gene Name
trafficking protein particle complex 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321102.1 Intron NP_001308031.1
NM_016030.5 Intron NP_057114.5
XM_011510350.2 Intron XP_011508652.1
XM_011510352.2 Intron XP_011508654.1
XM_011510353.2 Intron XP_011508655.1
XM_011510354.2 Intron XP_011508656.1
XM_011510355.2 Intron XP_011508657.1
XM_011510356.1 Intron XP_011508658.1
XM_017004257.1 Intron XP_016859746.1
Gene
TSSC1
Gene Name
tumor suppressing subtransferable candidate 1
There are no transcripts associated with this gene.

View Full Product Details