Product Details

SNP ID
rs13252406
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:143830133 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACATTTAGAATTTAATACAGACAC[C/T]TCTACTTAAATAGGGCAGAATATTC
Phenotype
MIM: 615563 MIM: 604819
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIR6845 PubMed Links

Gene Details

Gene
MIR6845
Gene Name
microRNA 6845
There are no transcripts associated with this gene.

Gene
NRBP2
Gene Name
nuclear receptor binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178564.3 Intron NP_848659.2
XM_017013378.1 Intron XP_016868867.1
XM_017013379.1 Intron XP_016868868.1
XM_017013380.1 Intron XP_016868869.1
XM_017013381.1 Intron XP_016868870.1
XM_017013382.1 Intron XP_016868871.1
XM_017013383.1 Intron XP_016868872.1
Gene
PUF60
Gene Name
poly(U) binding splicing factor 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136033.2 Intron NP_001129505.1
NM_001271096.1 Intron NP_001258025.1
NM_001271097.1 Intron NP_001258026.1
NM_001271098.1 Intron NP_001258027.1
NM_001271099.1 Intron NP_001258028.1
NM_001271100.1 Intron NP_001258029.1
NM_014281.4 Intron NP_055096.2
NM_078480.2 Intron NP_510965.1
XM_011516929.2 Intron XP_011515231.1
XM_011516930.2 Intron XP_011515232.1
XM_017013234.1 Intron XP_016868723.1
XM_017013235.1 Intron XP_016868724.1
XM_017013236.1 Intron XP_016868725.1
XM_017013237.1 Intron XP_016868726.1
XM_017013238.1 Intron XP_016868727.1
XM_017013239.1 Intron XP_016868728.1
XM_017013240.1 Intron XP_016868729.1

View Full Product Details