The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029782.5 | NP_001024953.1 | 6 - 7 | 670 | 84 |
NM_001383682.1 | 6 - 7 | 675 | 84 | ||
GenBank mRNA | AY349017.1 | - | 6 - 7 | 692 | 84 |
AY349018.1 | - | 3 - 4 | 332 | 84 |