Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-91
Entrez Gene ID
181621
Gene Name
protein-coding
Gene Aliases
CELE_Y15E3A.1
Chromosome Location
Chr.X: 15309374 - 15314895 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029782.5 NP_001024953.1 8 - 9 1325 61
NM_001383682.1 8 - 9 1330 61
GenBank mRNA AY349017.1 - 8 - 9 1347 61
AY349018.1 - 5 - 6 987 61

View Full Product Details