Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sprr-1
Entrez Gene ID
3564903
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_R03A10.6
Chromosome Location
Chr.X: 15466300 - 15470802 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078154.6 NP_510555.2 8 - 9 1371 76

View Full Product Details