Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ent-2
Entrez Gene ID
181648
Gene Name
Equilibrative Nucleoside Transporter
Gene Aliases
CELE_K09A9.3
Chromosome Location
Chr.X: 15601067 - 15603775 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078170.6 NP_510571.1 5 - 6 1046 79

View Full Product Details