Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdh-9
Entrez Gene ID
186608
Gene Name
CaDHerin family
Gene Aliases
CELE_F59C12.1
Chromosome Location
Chr.X: 16595238 - 16602243 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078284.5 NP_510685.2 11 - 12 1400 80

View Full Product Details