Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sod-3
Entrez Gene ID
181748
Gene Name
Superoxide dismutase [Mn] 2, mitochondrial
Gene Aliases
CELE_C08A9.1
Chromosome Location
Chr.X: 17092718 - 17102558 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078363.8 NP_510764.1 2 - 3 272 91
GenBank mRNA X85790.1 - - 232 91

View Full Product Details