Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npp-13
Entrez Gene ID
171774
Gene Name
protein-coding
Gene Aliases
CELE_Y37E3.15
Chromosome Location
Chr.I: 2097346 - 2123443 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306422.3 NP_001293351.1 3 - 4 864 134
NM_170819.6 NP_740802.1 5 - 6 1261 134

View Full Product Details