Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npp-13
Entrez Gene ID
171774
Gene Name
protein-coding
Gene Aliases
CELE_Y37E3.15
Chromosome Location
Chr.I: 2097346 - 2123443 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306422.3 NP_001293351.1 5 - 6 1059 100
NM_170819.6 NP_740802.1 7 - 8 1456 100
NR_131352.1 7 - 8 1295 100

View Full Product Details