Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-6
Entrez Gene ID
172156
Gene Name
protein-coding
Gene Aliases
CELE_F56A3.3
Chromosome Location
Chr.I: 5176830 - 5182659 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374886.2 NP_001361963.1 6 - 7 1677 78
NM_001380120.2 NP_001367609.1 6 - 7 1683 78

View Full Product Details