Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aman-3
Entrez Gene ID
172178
Gene Name
protein-coding
Gene Aliases
CELE_F48C1.1
Chromosome Location
Chr.I: 5325549 - 5330230 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374889.1 NP_001361919.1 4 - 5 572 75
NM_001374890.2 NP_001362173.1 4 - 5 579 75

View Full Product Details