Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fip-6
Entrez Gene ID
172707
Gene Name
Fungus-Induced Protein
Gene Aliases
CELE_F53B6.9
Chromosome Location
Chr.I: 8945533 - 8946244 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170908.8 NP_740901.1 1 - 2 124 105

View Full Product Details