Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
catp-1
Entrez Gene ID
173307
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8A.12
Chromosome Location
Chr.I: 14436810 - 14451889 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129056.5 NP_001122528.1 13 - 14 2790 79
NM_001129057.3 NP_001122529.1 13 - 14 2789 79
GenBank mRNA AM419897.1 - 13 - 14 2779 79
AM419898.1 - 13 - 14 2779 79

View Full Product Details