Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hot-4
Entrez Gene ID
175881
Gene Name
protein-coding
Gene Aliases
CELE_T12A2.16
Chromosome Location
Chr.III: 6215607 - 6224444 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171155.6 NP_741179.1 7 - 8 1595 95

View Full Product Details