Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lim-8
Entrez Gene ID
175952
Gene Name
protein-coding
Gene Aliases
CELE_F28F5.3
Chromosome Location
Chr.III: 6802788 - 6816141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066082.5 NP_498483.1 10 - 11 1887 116
NM_066083.7 NP_498484.1 10 - 11 2120 116
NM_171173.4 NP_741201.1 13 - 14 2115 116

View Full Product Details