Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exos-9
Entrez Gene ID
176012
Gene Name
protein-coding
Gene Aliases
CELE_F37C12.13
Chromosome Location
Chr.III: 7196224 - 7232172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372617.1 NP_001360028.1 7 - 8 746 72
NM_171186.6 NP_741217.1 8 - 9 768 72

View Full Product Details