Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
set-23
Entrez Gene ID
176969
Gene Name
protein-coding
Gene Aliases
CELE_Y41D4B.12
Chromosome Location
Chr.IV: 1601654 - 1605609 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171270.2 NP_741320.1 1 - 2 604 71
NM_171271.6 NP_741321.1 2 - 3 1883 71

View Full Product Details