The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001380163.1 | NP_001367796.1 | 2 - 3 | 218 | 72 |
NM_001380164.2 | NP_001367795.1 | 2 - 3 | 232 | 72 |