Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-75
Entrez Gene ID
177165
Gene Name
protein-coding
Gene Aliases
CELE_W03D2.1
Chromosome Location
Chr.IV: 4072291 - 4090475 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171311.6 NP_741365.1 5 - 6 475 77
NM_171312.5 NP_741366.1 4 - 5 452 77
NM_171903.6 NP_741367.1 4 - 5 389 77
NR_131466.1 4 - 5 332 77

View Full Product Details